Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAPPC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534886 Supplier Invitrogen™ Supplier No. PA534886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: H1299, HCT116, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (90%), Xenopus laevis (91%), Chicken (95%), Rhesus Monkey (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TRAPPC3 is a component of the TRAPP complex, which is involved in tethering of transport vesicles to the cis-Golgi membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC3
Applications Western Blot
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TRAPPC3
Gene Accession No. O43617, O55013
Gene Alias 1110058K12Rik; AI835942; AU019197; Bet3; BET3 homolog; CDABP0066; trafficking protein particle complex 3; trafficking protein particle complex subunit 3; TRAPPC3
Gene Symbols TRAPPC3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TRAPPC3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27095, 27096
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.