Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRAPPC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550920
Description
TRAPPC3 Polyclonal Antibody detects endogenous levels of TRAPPC3 protein.
TRAPPC3 is a component of the TRAPP complex, which is involved in tethering of transport vesicles to the cis-Golgi membrane.
Specifications
TRAPPC3 | |
Polyclonal | |
Unconjugated | |
TRAPPC3 | |
1110058K12Rik; AI835942; AU019197; Bet3; BET3 homolog; CDABP0066; trafficking protein particle complex 3; trafficking protein particle complex subunit 3; TRAPPC3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27095, 27096, 362599 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43617, O55013, Q5U1Z2 | |
TRAPPC3 | |
Synthesized peptide derived from the C-terminal region of human BET3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction