Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAPPC4 Antibody (OTI2G8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274594H
Description
TRAPPC4 Monoclonal specifically detects TRAPPC4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TRAPPC4 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
TRAPPC4 | |
Full length recombinant protein of human TRAPPC4 (NP_057230) produced in HEK293T cells. | |
0.1 mL | |
51399.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2G8 | |
Western Blot, Immunohistochemistry | |
Hematopoietic stem/progenitor cell protein 172, PTD009, SBDNHSPC172, Synbindin, trafficking protein particle complex 4, trafficking protein particle complex subunit 4, TRS23, TRS23 homolog | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction