Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAPPC4 Monoclonal Antibody (2D5)
GREENER_CHOICE

Catalog No. PIMA521923
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521923 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521923 Supplier Invitrogen™ Supplier No. MA521923
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MAIFSVYVVN KAGGLIYQLD SYAPRAEAEK TFSYPLDLLL KLHDERVLVA FGQRDGIRVG HAVLAINGMD VNGRYTADGK EVLEYLGNPA NYPVSIRFGR PRLTSNEKLM LASMFHSLFA IGSQLSPEQG SSGIEMLETD TFKLHCYQTL TGIKFVVLAD PRQAGIDSLL RKIYEIYSDF ALKNPFYSLE MPIRCELFDQ NLKLALEVAE KAGTFGPGS.

TRAPPC4 is part of the multisubunit TRAPP (transport protein particle) complex and interacts with SDC2. It may play a role in vesicular transport from endoplasmic reticulum to Golgi. TRAPP proteins are involved in tethering during vesicle transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2D5
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Trappc4
Gene Accession No. Q9Y296
Gene Alias 1500017G03Rik; AI303617; CGI-104; Hematopoietic stem/progenitor cell protein 172; HSPC172; hypothetical protein LOC504391; PTD009; Sbd; Sbdn; Sdcbp2; synbindin; syndecan binding protein 2; trafficking protein particle complex 4; trafficking protein particle complex subunit 4; Trappc4; TRS23; TRS23 homolog; zgc:73260
Gene Symbols Trappc4
Host Species Mouse
Immunogen TRAPPC4 (AAH10866, 1 a.a. ~ 219 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51399
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.