Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAPPC4 Monoclonal Antibody (OTI1D10), TrueMAB™, OriGene
SDP

Catalog No. 502539622
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9622 30 μL
50-167-6015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9622 Supplier OriGene Supplier No. TA505559S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAPPC4 Monoclonal antibody specifically detects TRAPPC4 in Human samples. It is validated for Western Blot

Specifications

Antigen TRAPPC4
Applications Western Blot
Classification Monoclonal
Clone OTI1D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene BTLA
Gene Accession No. Q7Z6A9
Gene Alias 1500017G03Rik; AI303617; CGI-104; Hematopoietic stem/progenitor cell protein 172; HSPC172; hypothetical protein LOC504391; PTD009; Sbd; Sbdn; Sdcbp2; synbindin; syndecan binding protein 2; trafficking protein particle complex 4; trafficking protein particle complex subunit 4; Trappc4; TRS23; TRS23 homolog; zgc:73260
Host Species Mouse
Immunogen Full length human recombinant protein of human BTLA produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151888
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.