Learn More
Invitrogen™ TRAPPC4 Monoclonal Antibody (OTI2E1)

Description
TRAPPC4 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | TRAPPC4 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2E1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Trappc4 |
| Gene Accession No. | Q9Y296 |
| Gene Alias | 1500017G03Rik; AI303617; CGI-104; Hematopoietic stem/progenitor cell protein 172; HSPC172; hypothetical protein LOC504391; PTD009; Sbd; Sbdn; Sdcbp2; synbindin; syndecan binding protein 2; trafficking protein particle complex 4; trafficking protein particle complex subunit 4; Trappc4; TRS23; TRS23 homolog; zgc:73260 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.