Learn More
TRAPPC4 Mouse anti-Human, Clone: OTI1A3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505600

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TRAPPC4 is part of the multisubunit TRAPP (transport protein particle) complex and interacts with SDC2. It may play a role in vesicular transport from endoplasmic reticulum to Golgi. TRAPP proteins are involved in tethering during vesicle transport.Specifications
TRAPPC4 | |
Monoclonal | |
Unconjugated | |
TRAPPC4 | |
CGI-104, HSPC172, PTD009, SBDN, SYNBINDIN, TRS23 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51399 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y296 | |
TRAPPC4 | |
Full length human recombit protein of human TRAPPC4 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.