Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAPPC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583552
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583552 Supplier Invitrogen™ Supplier No. PA583552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HCYQTLTGIK FVVLADPRQA GIDSLLRKIY EIYSDFALKN PFYSLEMPIR CELFDQNLKL ALEVAEKAGT Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

TRAPPC4 is part of the multisubunit TRAPP (transport protein particle) complex and interacts with SDC2. It may play a role in vesicular transport from endoplasmic reticulum to Golgi. TRAPP proteins are involved in tethering during vesicle transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Trappc4
Gene Accession No. Q69BT7, Q9ES56, Q9Y296
Gene Alias 1500017G03Rik; AI303617; CGI-104; Hematopoietic stem/progenitor cell protein 172; HSPC172; hypothetical protein LOC504391; PTD009; Sbd; Sbdn; Sdcbp2; synbindin; syndecan binding protein 2; trafficking protein particle complex 4; trafficking protein particle complex subunit 4; Trappc4; TRS23; TRS23 homolog; zgc:73260
Gene Symbols Trappc4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRAPPC4. Recombinant protein control fragment (Product #RP-97554).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 367073, 51399, 60409
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.