Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAPPC5 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318481G
Description
TRAPPC5 Polyclonal antibody specifically detects TRAPPC5 in Human, Mouse samples. It is validated for Western BlotSpecifications
TRAPPC5 | |
Polyclonal | |
Western Blot | |
MGC52424, trafficking protein particle complex 5, trafficking protein particle complex subunit 5, TRS31 | |
Recognizes region between residues Between between 138 and 188.. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
126003 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction