Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAPPC6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120428 Supplier Invitrogen™ Supplier No. PA5120428
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse kidney, Rat kidney, Rat liver. The target is usually found in the following locations: Endoplasmic reticulum, Golgi apparatus, cis-Golgi network. Immunogen sequence: MADTVLFEFL HTEMVAELWA HDPDPGPGVS AGLRGEEAGA TKGQKMSLSV LEGMGFRVGQ ALGERLPRET LAFREELDVL KFLCKDLWVA VFQKQMDSLR TNHQGTYVLQ DNSFPLLLPM ASGLQYLEEA PKFLAFTCGL LRGALYTLGI ESVVTASVAA LPVCKFQVVI PKS.

TRAPPC6A is a subunit of the TRAPP I and II complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC6A
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TRAPPC6A
Gene Accession No. O75865, Q78XR0
Gene Alias HSPC289; trafficking protein particle complex 6A; trafficking protein particle complex subunit 6A; TRAPP complex subunit 6A; TRAPPC6A; TRAPPC6Adelta29-42; TRS33
Gene Symbols TRAPPC6A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human TRAPPC6A (NP_077013.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67091, 680465, 79090
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.