Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAPPC6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561627
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561627 Supplier Invitrogen™ Supplier No. PA561627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MADEALFLLL HNEMVSGVYK SAEQGEVENG RCITKLE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 100%.

TRAPPC6B is a component of TRAPP complexes, which are tethering complexes involved in vesicle transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC6B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRAPPC6B
Gene Accession No. Q86SZ2
Gene Alias 5830498C14Rik; C79212; RGD1309325; TPC6; trafficking protein particle complex 6b; trafficking protein particle complex subunit 6b; Trafficking protein particle complex subunit 6B-like protein; TRAPP complex subunit 6B; Trappc6b; Unknown (protein for MGC:133693); zgc:103464
Gene Symbols TRAPPC6B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRAPPC6B. Recombinant protein control fragment (Product #RP-100490).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122553
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.