Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106442
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106442 Supplier Invitrogen™ Supplier No. PA5106442
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TRAPPC9.

NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIBP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TRAPPC9
Gene Accession No. Q3U0M1, Q96Q05
Gene Alias 1810044A24Rik; 2900005P22Rik; 4632408O18Rik; hypothetical protein LOC533451; Ibp; IKBKBBP; IKK2 binding protein; Kiaa1882; mKIAA1882; MRT13; Nibp; NIK- and IKBKB-binding protein; NIK and IKK(beta) binding protein; NIK and IKK{beta} binding protein; NIK and IKK-beta binding protein; RGD1309461; T1; trafficking protein particle complex 9; trafficking protein particle complex subunit 9; TRAPP 120 kDa subunit; TRAPPC9; TRS120; TRS130; tularik gene 1 protein
Gene Symbols TRAPPC9
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRAPPC9(Accession Q96Q05), corresponding to amino acid residues F788-N838.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100334971, 315059, 76510, 83696
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.