Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trappin-2/Elafin/Skalp Antibody (101), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290071APC
Description
Trappin-2/Elafin/Skalp Monoclonal antibody specifically detects Trappin-2/Elafin/Skalp in Human samples. It is validated for ELISASpecifications
Trappin-2/Elafin/Skalp | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
101 | |
ELISA | |
cementoin, Elastase-specific inhibitor, ESIWAP four-disulfide core domain protein 14, Peptidase inhibitor 3, peptidase inhibitor 3, skin-derived, PI-3, pre-elafin, protease inhibitor 3, skin-derived (SKALP), Protease inhibitor WAP3, SKALPELAFIN, Skin-derived antileukoproteinase, trappin-2, WAP four-disulfide core domain 14, WAP3MGC13613, WFDC14elafin | |
Rabbit immunized with purified, recombinant Human Trappin-2/Elafin/Skalp (Accession#: NP_002629.1, Met1-Gln117) | |
0.1 mL | |
Cancer, Immunology | |
5266 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction