Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trastuzumab Antibody (2261E), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB95471MFV610
Description
Trastuzumab Monoclonal antibody specifically detects Trastuzumab in Broad Species Reactivity samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
| Trastuzumab | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Full-length Trastuzumab | |
| RUO | |
| Broad Species Reactivity | |
| IgG |
| Western Blot, Flow Cytometry, ELISA | |
| 2261E | |
| Western Blot, Flow Cytometry, ELISA | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction