Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti- Trastuzumab (Anti-Idiotype) Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB95471G100UG
This item is not returnable.
View return policy
Description
Trastuzumab Monoclonal specifically detects Trastuzumab in Multi-Species samples. It is validated for Flow Cytometry.Specifications
| Trastuzumab | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide | |
| Full-length Trastuzumab | |
| 100 μg | |
| Detects Trastuzumab in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Adalimumab, Tositumomab, Brentuximab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG |
| Flow Cytometry | |
| 2261E | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Multi-species | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction