Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728620 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8620 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8620 Supplier Proteintech Group Inc Supplier No. 132791AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM (T cell receptor-interacting molecule), also known as TRAT1 (T cell receptor associated transmembrane adaptor 1) is a 30 kDa protein expressed by T cells as a cystein-linked homodimer. It associates with TCR-CD3-zeta-chain complex and becomes phosphorylated by Src-family kinases. TRIM is potentially involved in negative regulation of TCR-mediated signaling, but its role remains unclear.

Specifications

Antigen TRAT1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TRAT1
Gene Accession No. Q3UU67, Q6PIZ9
Gene Alias HSPC062, pp29/30, TCRIM, TRAT1, TRIM
Gene Symbols TRAT1
Host Species Rabbit
Immunogen TRAT1 Fusion Protein Ag4023
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498075, 50852, 77647
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.