Learn More
Eisco™ Travertine Sedimentary Rock Specimen
SCI_ED
Catalog No. S27611 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27611 12/Pk.
S27589 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27611 Supplier Eisco™ Supplier No. ESNG0082PK12
Add to Cart
Add to Cart
Travertine is a carbonate rock formed from gushing hot springs. The airy, porous material indicates an abundance of air bubbles moving through the rock as it solidified. Travertine is unique in that it forms as a result of rapidly dropping pH levels.
  • Correlates well to Next Generation Science Standards Evidence
  • Ideal for teaching students about rounding and sorting, as well as for identification & classification exercises
  • Useful in lessons on sedimentary petrology and geothermal systems
Specifications
Product Type Sedimentary Rock
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg