Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRBP Antibody (PCRP-TARBP2-1E5), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308235AF532
Description
TRBP Monoclonal antibody specifically detects TRBP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
TRBP | |
Monoclonal | |
Alexa Fluor 532 | |
LOQS, RISC-loading complex subunit TARBP2, TAR (HIV) RNA binding protein 2, TAR (HIV) RNA-binding protein 2, TAR (HIV) RNA-binding protein TRBP1, TAR (HIV-1) RNA binding protein 2, TAR RNA binding protein 2, TAR RNA-binding protein 2, trans-activation responsive RNA-binding protein, Trans-activation-responsive RNA-binding protein, TRBP, TRBP1, TRBP2 | |
Recombinant full-length human TRBP protein (Uniprot: Q15633) | |
100 μg | |
DNA replication Transcription Translation and Splicing | |
6895 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-TARBP2-1E5 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction