Learn More
Invitrogen™ TRBP Monoclonal Antibody (OTI7C1)

Description
TRBP Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | TRBP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI7C1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TARBP2 |
| Gene Accession No. | Q15633 |
| Gene Alias | fj51e12; HAMAP-Rule:MF_03034}; LOQS; Prbp; PRM-1 RNA-binding protein; protamine-1 RNA-binding protein; RISC-loading complex subunit tarbp2; RISC-loading complex subunit TARBP2 {ECO:0000255; TAR (HIV) RNA binding protein 2; TAR (HIV) RNA-binding protein 2; TAR (HIV) RNA-binding protein TRBP1; TAR (HIV-1) RNA binding protein 2; TAR RNA binding protein 2; TAR RNA-binding protein 2; TARBP2; TARBP2 subunit of RISC loading complex; TARBP2, RISC loading complex RNA binding subunit; trans-activation responsive RNA-binding protein; trans-activation-responsive RNA-binding protein; TRBP; TRBP1; TRBP2; wu:fj51e12; zgc:63778 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.