Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584500 Supplier Invitrogen™ Supplier No. PA584500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSVVLTRSPP MELQPPVSPQ QSECNPVGAL QELVVQKGWR LPEYTVTQES GPAHRKEFTM TCRVERFIEI GSG Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 95%.

TRBP belongs to the family of dsRNA-binding proteins. It consists of highly conserved 2 dsRNA-binding domains and C-terminal basic region. It is a unique cellular protein capable of binding to HIV-TAR RNA and stimulates expression of HIV-1 long terminal repeats. It also inhibits the host antiviral and anti-proliferative mechanisms by directly binding to IFN-regulated dsRNA-dependent protein kinase (PKR). TRBP associates with Ago2 protein and forms an integral component of Dicer-containing complex. It has a role in micro RNA processing and in RISC assembly. Other functions of TRBP include promoting cellular growth and tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRBP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TARBP2
Gene Accession No. Q15633
Gene Alias fj51e12; HAMAP-Rule:MF_03034}; LOQS; Prbp; PRM-1 RNA-binding protein; protamine-1 RNA-binding protein; RISC-loading complex subunit tarbp2; RISC-loading complex subunit TARBP2 {ECO:0000255; TAR (HIV) RNA binding protein 2; TAR (HIV) RNA-binding protein 2; TAR (HIV) RNA-binding protein TRBP1; TAR (HIV-1) RNA binding protein 2; TAR RNA binding protein 2; TAR RNA-binding protein 2; TARBP2; TARBP2 subunit of RISC loading complex; TARBP2, RISC loading complex RNA binding subunit; trans-activation responsive RNA-binding protein; trans-activation-responsive RNA-binding protein; TRBP; TRBP1; TRBP2; wu:fj51e12; zgc:63778
Gene Symbols TARBP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TARBP2. Recombinant protein control fragment (Product #RP-104214).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6895
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.