Learn More
Invitrogen™ TRBP Recombinant Rabbit Monoclonal Antibody (24GB2425)
Rabbit Recombinant Monoclonal Antibody
$189.00 - $539.00
Specifications
Antigen | TRBP |
---|---|
Clone | 24GB2425 |
Content And Storage | -20°C |
Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
Classification | Recombinant Monoclonal |
Description
TRBP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TRBP belongs to the family of dsRNA-binding proteins. It consists of highly conserved 2 dsRNA-binding domains and C-terminal basic region. It is a unique cellular protein capable of binding to HIV-TAR RNA and stimulates expression of HIV-1 long terminal repeats. It also inhibits the host antiviral and anti-proliferative mechanisms by directly binding to IFN-regulated dsRNA-dependent protein kinase (PKR). TRBP associates with Ago2 protein and forms an integral component of Dicer-containing complex. It has a role in micro RNA processing and in RISC assembly. Other functions of TRBP include promoting cellular growth and tumorigenesis.Specifications
TRBP | |
-20°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat | |
TARBP2 | |
fj51e12; HAMAP-Rule:MF_03034}; LOQS; Prbp; PRM-1 RNA-binding protein; protamine-1 RNA-binding protein; RISC-loading complex subunit tarbp2; RISC-loading complex subunit TARBP2 {ECO:0000255; TAR (HIV) RNA binding protein 2; TAR (HIV) RNA-binding protein 2; TAR (HIV) RNA-binding protein TRBP1; TAR (HIV-1) RNA binding protein 2; TAR RNA binding protein 2; TAR RNA-binding protein 2; TARBP2; TARBP2 subunit of RISC loading complex; TARBP2, RISC loading complex RNA binding subunit; trans-activation responsive RNA-binding protein; trans-activation-responsive RNA-binding protein; TRBP; TRBP1; TRBP2; wu:fj51e12; zgc:63778 | |
TARBP2 | |
Primary | |
Affinity Chromatography |
24GB2425 | |
Flow Cytometry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P97473, Q15633, Q3SWU0 | |
21357, 363006, 6895 | |
A synthesized peptide derived from human TRBP (aa 230-280). | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.