Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555130
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555130 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555130 Supplier Invitrogen™ Supplier No. PA555130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNELNSVSDL DRCHLYLMVL TELINLHLKV GWKRGNPIWR VISLLKNASI QHLQEMDSGQ EPTVGSQIQR VVSMAALAMV CEAIDQKPEL QLDSLHAG Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 59%.

The human trans-activation response (TAR) RNA binding protein 1 (TRBP1) was initially identified as a protein that binds to the HIV-1 TAR RNA and activates the long terminal repeat (LTR) expression in the absence and presence of the viral trans-activator Tat. This binding is enhanced by the presence of co-factors such as elongation factor 1-a (EF-1-a), polypyrimidine tract-binding protein (PTB), and the chaperonin-like protein stimulator of TAR RNA-binding proteins (SRB). TRBP1 may act to disengage RNA polymerase II from TAR during transcriptional elongation if the RNA polymerase stalls at the TAR during transcriptional elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRBP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TARBP1
Gene Accession No. Q13395
Gene Alias Gm17296; Gm179; predicted gene, 17296; probable methyltransferase TARBP1; RP5-827C21.5; TAR (HIV) RNA binding protein 1; TAR (HIV) RNA-binding protein 1; TAR (HIV-1) RNA binding protein 1; TAR RNA loop binding protein; TAR RNA-binding protein 1; TAR RNA-binding protein of 185 kDa; Tarbp1; TRM3; tRNA methyltransferase 3 homolog; TRP185; TRP-185
Gene Symbols TARBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRBP1. Recombinant protein control fragment (Product #RP-91765).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.