Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TREH Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522076

Catalog No. PIPA522076


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse stomach. Predicted reactivity: Mouse (80%), Rat (80%), Rabbit (82%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes an enzyme that hydrolyses trehalose, a disaccharide formed from two glucose molecules found mainly in fungi, plants, and insects. A partial duplication of this gene is located adjacent to this locus on chromosome 11.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TREH
Polyclonal
Unconjugated
TREH
2210412M19Rik; Alpha,alpha-trehalase; alpha,alpha-trehalose glucohydrolase; LOW QUALITY PROTEIN: trehalase; trehalase; si:ch211-147p17.2; TRE; TREA; TREH; Trehalase; trehalase (brush-border membrane glycoprotein)
Rabbit
Antigen Affinity Chromatography
RUO
11181, 58866
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O43280, Q9JLT2
TREH
Recombinant fragment corresponding to a region within amino acids 7 and 285 of Trehalase (Uniprot ID#O43280).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.