Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trehalase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984013
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4013 Supplier Bioss Supplier No. BS16712R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes an enzyme that hydrolyses trehalose, a disaccharide formed from two glucose molecules found mainly in fungi, plants, and insects. A partial duplication of this gene is located adjacent to this locus on chromosome 11.

Specifications

Antigen Trehalase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TREH
Gene Accession No. O43280
Gene Alias 2210412M19Rik; Alpha,alpha-trehalase; Alpha,alpha-trehalose glucohydrolase; BOS_14553; si:ch211-147p17.2; TRE; TREA; Treh; Trehalase; trehalase (brush-border membrane glycoprotein)
Gene Symbols TREH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Trehalase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11181
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.