Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Trehalose-6-phosphate phosphatase Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121447
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.
Removes the phosphate from trehalose 6-phosphate (Tre6P) to produce free trehalose. Also catalyzes the dephosphorylation of glucose-6-phosphate (Glu6P) and 2-deoxyglucose-6-phosphate (2dGlu6P).
Specifications
| Trehalose-6-phosphate phosphatase | |
| Polyclonal | |
| Unconjugated | |
| otsB | |
| otsB | |
| Recombinant Escherichia coli Trehalose-6-phosphate phosphatase protein (1-266aa). | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3.02 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P31678 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 946406 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction