Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREK 1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11041535AF488
Description
TREK 1 Polyclonal specifically detects TREK 1 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry (Negative), ICC/IF (Negative).Specifications
TREK 1 | |
Polyclonal | |
Western Blot, Immunohistochemistry (Negative), ICC/IF (Negative) | |
O95069 | |
KCNK2 | |
A synthetic peptide made to an N-terminal region of human TREK 1 (within residues 1-100). [Swiss-Prot# Q9NRT2] | |
0.1 mL | |
Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
3776 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MGC126742, MGC126744, potassium channel, subfamily K, member 2, subfamily K, member 2, tandem-pore-domain potassium channel TREK-1, TREK-1 K(+) channel subunit, TWIK-related potassium channel 1, two-pore potassium channel 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Bovine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction