Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREK 1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $499.50

Specifications

Antigen TREK 1
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Immunohistochemistry (Negative), ICC/IF (Negative)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11041535T
SDP
View Documents
Novus Biologicals
NB11041535SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB11041535
SDP
View Documents
Novus Biologicals
NB11041535
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

TREK 1 Polyclonal specifically detects TREK 1 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry (Negative), ICC/IF (Negative).
Specifications

Specifications

TREK 1
Western Blot 1:1000, Immunohistochemistry (Negative), ICC/IF (Negative)
Polyclonal
Rabbit
Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
MGC126742, MGC126744, potassium channel, subfamily K, member 2, subfamily K, member 2, tandem-pore-domain potassium channel TREK-1, TREK-1 K(+) channel subunit, TWIK-related potassium channel 1, two-pore potassium channel 1
KCNK2
IgG
Affinity Purified
Human, mouse, and cow. 88% sequence identity with rat protein.
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry
Unconjugated
RUO
Human, Mouse, Bovine
O95069
3776
A synthetic peptide made to an N-terminal region of human TREK 1 (within residues 1-100). [Swiss-Prot# Q9NRT2]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.