Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM-1 (CD354) Mouse anti-Human, Brilliant Violet 421, Clone: 6B1, BD Optibuild™

Catalog No. BDB743738 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743738 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743738 Supplier BD Biosciences Supplier No. 743738
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 6B1monoclonal antibody specifically binds to TREM-1 (Triggering Receptor Expressed on Myeloid Cells 1 or Triggering Receptor Expressed on Monocytes 1) which is also known as CD354. CD354 is a type I transmembrane glycoprotein that belongs to the immunoglobulin superfamily. CD354 is highly expressed by neutrophils and monocytes. It is also reportedly expressed at lower levels by a variety of cells including subsets of T and B cells, NK cells and dendritic cells. Its expression is upregulated on leucocytes in response to microbial lipopolysaccharide, eg, during microbial infections or sepsis. CD354 activation triggers the release of proinflammatory cytokines and chemokines. A soluble form of CD354 has been described that may play a protective role in some infectious disease states.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREM-1 (CD354)
Classification Monoclonal
Clone 6B1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TREM1, Triggering receptor expressed on myeloid cells 1
Host Species Mouse
Immunogen Human Trem-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.