Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM-2 ectodomain Rat anti-Human, Mouse, Clone: 5F4, MilliporeSigma™

Catalog No. MABN232125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN232125 25 μg
MABN2321100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN232125 Supplier MilliporeSigma Supplier No. MABN232125UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TREM-2 ectodomain Monoclonal antibody specifically detects TREM-2 ectodomain in Human, Mouse samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREM-2 ectodomain
Applications Western Blot
Classification Monoclonal
Clone 5F4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rat
Immunogen His-tagged recombinant extracellular domain of murine TREM-2.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.