Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TREM-2, Mouse, Unlabeled, Clone: 10H4.2,

Catalog No. MABN1534MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1534MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1534MI Supplier MilliporeSigma™ Supplier No. MABN1534
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protein S100-A4 (UniProt P26447; also known as Calvasculin, Metastasin, Placental calcium-binding protein, Protein Mts1, S100 calcium-binding protein A4) is encoded by the S100A4 (also known as CAPL, MTS1, P9KA, PEL98) gene (Gene ID 6275) in human. S100A4 belongs to a family of low moleclar weight proteins characterized by two calcium-binding helix-loop-helix EF-hand domains (a.a. 12-47 & 50-85 of human S100A4). S100 proteins exist as non-covalent homodimers with cell-specific expression patterns. S100A4 associates with cytoskeletal proteins, such as myosin and F-actin, as well as other actin-binding proteins such as Talin and integrins, and is involved in controlling not just shape and motility of normal cells, but also plays a role in cancer cells invasion/metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREM-2
Applications Western Blot
Classification Monoclonal
Clone 10H4.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene TREM2
Gene Accession No. Q9NZC2
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to a cytoplasmic domain sequence of human TREM-2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_061838
Test Specificity Clone 10H4.2 recognizes a cytoplasmic domain epitope that is not present in the spliced isoforms 2&3 reported by UniProt (Q9NZC2).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.