Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM-2 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABN1471MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1471MI 100 μg
ABN1471-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN1471MI Supplier MilliporeSigma Supplier No. ABN1471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-TREM-2 is a highly specific rabbit polyclonal antibody that targets Triggering receptor expressed on myeloid cells 2 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREM-2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols TREM2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal region of human TREM-2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_061838
Test Specificity This polyclonal antibody antibody detects Triggering receptor expressed on myeloid cells 2 in several regions of human brain. It targets an epitope within 13 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.