Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM-2 Rat anti-Mouse, Unconjugated, Clone: 14D1, MilliporeSigma™

Catalog No. MABN23202MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN23202MI 25 μg
MABN23201MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN23202MI Supplier MilliporeSigma Supplier No. MABN232025UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TREM-2 Monoclonal antibody specifically detects TREM-2 in Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREM-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 14D1
Conjugate Unconjugated
Gene Accession No. NP_001259007
Gene Alias Triggering receptor expressed on myeloid cells 2;Triggering receptor expressed on monocytes 2
Gene Symbols Trem2, Trem2a, Trem2c
Host Species Rat
Immunogen A linear peptide corresponding to 20 amino acids from the C-terminal region of murine Triggering receptor expressed on myeloid cells 2 (TREM-2).
Molecular Weight of Antigen 24.53 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 14D1 is a rat monoclonal antibody that specifically detects murine Triggering receptor expressed on myeloid cells 2 (TREM-2). It targets an epitope with in 20 amino acids from the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.