Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM1 Antibody (6B1), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP321495AF750
Description
TREM1 Monoclonal antibody specifically detects TREM1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
TREM1 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
6B1 | |
Western Blot, Flow Cytometry, ELISA | |
CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
Recombinant human TREM1 fused with IgG | |
0.1 mL | |
Signal Transduction | |
54210 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction