Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM1 Antibody (MM0583-9T3), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211977V
Description
TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TREM1 | |
Monoclonal | |
DyLight 405 | |
Q9NP99 | |
TREM1 | |
Human recombinant TREM1 (extracellular domain) | |
0.1 mL | |
Signal Transduction | |
54210 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0583-9T3 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction