Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TREM1 Monoclonal Antibody (CL7111)
GREENER_CHOICE

Catalog No. PIMA535040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535040 Supplier Invitrogen™ Supplier No. MA535040
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: KLTEEKYELK EGQTLDVK Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 71%.

TREM1 (triggering receptor expressed on myeloid cells 1) is a receptor belonging to the Ig superfamily that amplifies neutrophil and monocyte-mediated inflammatory responses triggered by bacterial and fungal infections by stimulating release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. It is thought that TREM1 other related TREM proteins such as TREM2 act to modulate the inflammatory response. TREM1 has also been linked to inflammatory bowel disease resulting in enhanced production of TNF, IL-6, IL-8, and MCP-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREM1
Applications Western Blot
Classification Monoclonal
Clone CL7111
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TREM1
Gene Accession No. Q9NP99
Gene Alias CD354; sCD354; soluble CD354; soluble TREM 1; soluble TREM1; sTREM 1; sTREM1; TREM; Trem1; TREM-1; triggering receptor expressed in monocytes 1; triggering receptor expressed on monocytes 1; Triggering receptor expressed on myeloid cells; Triggering receptor expressed on myeloid cells 1
Gene Symbols TREM1
Host Species Mouse
Immunogen Synthetic Peptide corresponding to Human TREM1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54210
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.