Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM2 Antibody (08), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP321453AF647
Description
TREM2 Monoclonal antibody specifically detects TREM2 in Human samples. It is validated for ELISA, ELISA (Matched Antibody Pair)Specifications
TREM2 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
08 | |
ELISA, ELISA Detection (Matched Antibody Pair) | |
Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
Purified, recombinant Human TREM2 (NBP3-21453PEP; NP_061838.1; Met1-Ser174) | |
0.1 mL | |
Alzheimers Research, Immunology, Inflammation, Innate Immunity, Neuroscience | |
54209 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction