Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM2 Antibody (237920R), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB17291RX
Description
TREM2 Monoclonal antibody specifically detects TREM2 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
TREM2 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
237920R | |
Flow Cytometry | |
Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TREM2, Met1-Pro168, Accession # Q99NH8 | |
0.1 mL | |
Alzheimers Research, Immunology, Inflammation, Innate Immunity, Neuroscience | |
54209 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction