Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TREM2 Monoclonal Antibody (78.18)
GREENER_CHOICE

Catalog No. PIMA528224
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528224 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528224 Supplier Invitrogen™ Supplier No. MA528224
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Rat anti Mouse TREM-2 antibody, clone 78.18 recognizes triggering receptor expressed on myeloid cells 2 (TREM-2), an activating receptor of the Ig superfamily.

TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREM2
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 78.18
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Trem2
Gene Accession No. Q99NH8
Gene Alias OTTHUMP00000039771; soluble TREM 2; soluble TREM2; sTREM 2; sTREM2; TREM; Trem2; TREM-2; Trem2a; Trem2b; Trem2c; Triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells 2; triggering receptor expressed on myeloid cells 2a; triggering receptor expressed on myeloid cells 2b; triggering receptor expressed on myeloid cells 2c
Gene Symbols Trem2
Host Species Rat
Immunogen Mouse TREM-2 Fc fusion protein.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83433
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.