Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM2 Polyclonal Antibody

TREM2 Polyclonal Antibody
Supplier: Thermo Scientific PA518764
Description
TREM2 Polyclonal Antibody for ICC/IF
Specifications
| TREM2 | |
| Polyclonal | |
| Unconjugated | |
| Trem2 | |
| OTTHUMP00000039771; soluble TREM 2; soluble TREM2; sTREM 2; sTREM2; TREM; Trem2; TREM-2; Trem2a; Trem2b; Trem2c; Triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells 2; triggering receptor expressed on myeloid cells 2a; triggering receptor expressed on myeloid cells 2b; triggering receptor expressed on myeloid cells 2c | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 83433 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q99NH8 | |
| Trem2 | |
| Synthetic peptide sequence (QVEHSTSRNQET) corresponding to the internal amino acids of Trem2 (aa 154-166). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction