Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM2b Antibody (RM0139-5J46), mFluor Violet 500 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122515MFV500
Description
TREM2b Monoclonal antibody specifically detects TREM2b in Mouse samples. It is validated for Flow CytometrySpecifications
TREM2b | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
54209 | |
Store at 4C in the dark. | |
IgG2 |
Flow Cytometry | |
RM0139-5J46 | |
Flow Cytometry | |
Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
Mouse recombinant TREM2b (extracellular domain) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction