Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tremelimumab Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541844
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541844 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541844 Supplier Invitrogen™ Supplier No. MA541844
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Tremelimumab is under investigation for the treatment of Mesothelioma, Liver Cancer, Liver Neoplasms, Liver Cell Carcinoma, and Hepatocellular Carcinoma. Tremelimumab has been investigated in Part C: Malignant Mesothelioma and Part A and B: Advanced Solid Malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tremelimumab
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1.21 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias CP-675; CP-675206 clone 11.2.1
Host Species Human
Immunogen Human CTLA4/CD152.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.