Learn More
Invitrogen™ TREML1 Monoclonal Antibody (268420)

Description
In direct ELISAs, no cross-reactivity with recombinant mouse TREML1 or recombinant human TREML2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | TREML1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 268420 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | TREML1 |
| Gene Accession No. | Q86YW5 |
| Gene Alias | 5430401J17Rik; dJ238O23.3; GLTL1825; MGC119173; OTTHUMP00000017858; PRO3438; RGD1565175; Tlt1; TLT-1; TREM; TREML1; trem-like transcript 1 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 1; triggering receptor expressed on myeloid cells-like 1; triggering receptor expressed on myeloid cells-like 1c; triggering receptor expressed on myeloid cells-like 1d; triggering receptor expressed on myeloid cells-like 1e; triggering receptor expressed on myeloid cells-like protein 1; UNQ1825/PRO3438 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.