Learn More
Invitrogen™ TREML1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587881
Description
Immunogen sequence: QGIVGSLPEV LQAPVGSSIL VQCHYRLQDV KAQKVWCRFL PEGCQPLVSS AVDRRAPAGR RTFLTDLGGG LLQVEMVTLQ EEDAGEYGCM VDGARGPQIL HRVSLNILPP EEEEETHKIG SLAENAFSDP AGSANPLEPS QDEKSIP; Positive Samples: Mouse liver, Mouse lung, Rat liver; Cellular Location: Cell membrane, Cytoplasm, Single-pass type I membrane protein.
TREML1 is located in a gene cluster on chromosome 6 with the single Ig variable domain activating receptors TREM1 and TREM2, but it has distinct structural and functional properties. TREML1 enhances calcium signaling in an SHP2 -dependent manner.
Specifications
TREML1 | |
Polyclonal | |
Unconjugated | |
TREML1 | |
5430401J17Rik; dJ238O23.3; GLTL1825; MGC119173; OTTHUMP00000017858; PRO3438; RGD1565175; Tlt1; TLT-1; TREM; TREML1; trem-like transcript 1 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 1; triggering receptor expressed on myeloid cells-like 1; triggering receptor expressed on myeloid cells-like 1c; triggering receptor expressed on myeloid cells-like 1d; triggering receptor expressed on myeloid cells-like 1e; triggering receptor expressed on myeloid cells-like protein 1; UNQ1825/PRO3438 | |
Rabbit | |
Affinity chromatography | |
RUO | |
501096, 71326 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K558 | |
TREML1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 16-162 of human TREML1 (NP_8354681). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.