Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TREML1 Recombinant Rabbit Monoclonal Antibody (027)
GREENER_CHOICE

Catalog No. PIMA530972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530972 Supplier Invitrogen™ Supplier No. MA530972
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human TREML1.

TREML1 is located in a gene cluster on chromosome 6 with the single Ig variable domain activating receptors TREM1 and TREM2, but it has distinct structural and functional properties. TREML1 enhances calcium signaling in an SHP2 -dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREML1
Applications ELISA
Classification Recombinant Monoclonal
Clone 027
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TREML1
Gene Accession No. Q86YW5
Gene Alias 5430401J17Rik; dJ238O23.3; GLTL1825; MGC119173; OTTHUMP00000017858; PRO3438; RGD1565175; Tlt1; TLT-1; TREM; TREML1; trem-like transcript 1 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 1; triggering receptor expressed on myeloid cells-like 1; triggering receptor expressed on myeloid cells-like 1c; triggering receptor expressed on myeloid cells-like 1d; triggering receptor expressed on myeloid cells-like 1e; triggering receptor expressed on myeloid cells-like protein 1; UNQ1825/PRO3438
Gene Symbols TREML1
Host Species Rabbit
Immunogen Recombinant Human TREML1 protein (Met1-Pro162).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340205
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.