Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREML1/TLT-1 Antibody (027), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290024JF549
Description
TREML1/TLT-1 Monoclonal antibody specifically detects TREML1/TLT-1 in Human samples. It is validated for ELISASpecifications
TREML1/TLT-1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
340205 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
027 | |
ELISA | |
dJ238O23.3, GLTL1825, PRO3438, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1 | |
Rabbit immunized with purified, recombinant Human TREML1/TLT-1 (Uniprot#: Q86YW5-1, Met1-Pro162) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction