Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREML1/TLT-1 Antibody (268420), DyLight 755, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2394Z
Description
TREML1/TLT-1 Monoclonal antibody specifically detects TREML1/TLT-1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
TREML1/TLT-1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
340205 | |
Store at 4°C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry, CyTOF | |
268420 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
dJ238O23.3, GLTL1825, PRO3438, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1 | |
Mouse myeloma cell line NS0-derived recombinant human TREML1/TLT-1, Gln16-Ser160, Accession # Q86YW5 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction