Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TREML2 Monoclonal Antibody (656906)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA524295
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
TREML2 is located in a gene cluster on chromosome 6 with the single Ig variable (IgV) domain activating receptors TREM1 and TREM2, but it has distinct structural and functional properties.
Specifications
TREML2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q2LA85 | |
TREML2 | |
CHO-derived recombinant mouse TREML2/TLT-2 His25-Ala270. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
656906 | |
Unconjugated | |
TREML2 | |
AW049306; C6orf76; dJ238O23.1; FLJ13693; Gm750; MGC149715; MGC149716; Tlt2; TLT-2; TREM; Treml2; Trem-like transcript 2 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 2; triggering receptor expressed on myeloid cells-like 2; triggering receptor expressed on myeloid cells-like 2a dominant; triggering receptor expressed on myeloid cells-like protein 2; UNQ6268/PRO20473 | |
Rat | |
Protein A/G | |
RUO | |
328833 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction