Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TREML2 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547833
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) TREML1, rmTREML4, rmTREM1, rmTREM2b, rmTREM3, rmTREM4, and recombinant human TREM2 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
TREML2 is located in a gene cluster on chromosome 6 with the single Ig variable (IgV) domain activating receptors TREM1 and TREM2, but it has distinct structural and functional properties.
Specifications
TREML2 | |
Polyclonal | |
Unconjugated | |
TREML2 | |
AW049306; C6orf76; dJ238O23.1; FLJ13693; Gm750; MGC149715; MGC149716; Tlt2; TLT-2; TREM; Treml2; Trem-like transcript 2 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 2; triggering receptor expressed on myeloid cells-like 2; triggering receptor expressed on myeloid cells-like 2a dominant; triggering receptor expressed on myeloid cells-like protein 2; UNQ6268/PRO20473 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
328833 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q2LA85 | |
TREML2 | |
CHO-derived recombinant mouse TREML2/TLT-2 His25-Ala270. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction