Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TREML2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107295
Description
Sequence homology: Dog: 79%; Human: 100%.
TREML2 is located in a gene cluster on chromosome 6 with the single Ig variable (IgV) domain activating receptors TREM1 and TREM2, but it has distinct structural and functional properties.
Specifications
| TREML2 | |
| Polyclonal | |
| Unconjugated | |
| TREML2 | |
| AW049306; C6orf76; dJ238O23.1; FLJ13693; Gm750; MGC149715; MGC149716; Tlt2; TLT-2; TREM; Treml2; Trem-like transcript 2 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 2; triggering receptor expressed on myeloid cells-like 2; triggering receptor expressed on myeloid cells-like 2a dominant; triggering receptor expressed on myeloid cells-like protein 2; UNQ6268/PRO20473 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79865 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q5T2D2 | |
| TREML2 | |
| Synthetic peptide directed towards the middle region of human TREML2 (aa 188-238). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction