Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREML2/TLT-2 Antibody (656906) [DyLight 350], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6707D
Description
TREML2/TLT-2 Monoclonal antibody specifically detects TREML2/TLT-2 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
TREML2/TLT-2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
79865 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
656906 | |
Flow Cytometry, CyTOF-ready | |
C6orf76, dJ238O23.1, FLJ13693, MGC149715, MGC149716, TLT2, TLT-2, trem-like transcript 2 protein, triggering receptor expressed on myeloid cells-like 2, triggering receptor expressed on myeloid cells-like protein 2, UNQ6268/PRO20473 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse TREML2/TLT-2, His25-Ala270, Accession # Q2LA85 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction